missing translation for 'onlineSavingsMsg'
Learn More

Methyl iso-Butyl Ketone (Certified ACS), Fisher Chemical™

Catalog No. p-163577
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
Packaging:
Glass Bottle
Steel Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M2131 1 L Glass Bottle
M2134 4 L Glass Bottle
M21320 20 L Steel Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M2131 Supplier Thermo Fisher Scientific Formerly Fairlawn Supplier No. M213 1
Only null left

Chemical Identifiers

CAS 108-10-1
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.161
MDL Number MFCD00008938
InChI Key NTIZESTWPVYFNL-UHFFFAOYSA-N
Synonym 4-methyl-2-pentanone, methyl isobutyl ketone, isopropylacetone, isobutyl methyl ketone, hexone, mibk, 2-pentanone, 4-methyl, 2-methyl-4-pentanone, 4-methyl-2-oxopentane, methylisobutylketon
PubChem CID 7909
ChEBI CHEBI:82344
IUPAC Name 4-methylpentan-2-one
SMILES CC(C)CC(=O)C

Specifications

Melting Point -84°C
Density 0.80g/cm³
Boiling Point 117°C
Quantity 1 L
Identification Pass Test
Packaging Glass Bottle
Physical Form Liquid
Residue after Evaporation 0.005% max.
Assay Percent Range ≥98.5 %
Titratable Acid 0.002mEq/g max.
Vapor Pressure 19.9mmHg at 25°C
Water 0.1% max.
Formula Weight 100.16
Percent Purity ≥98.5%
Grade Certified ACS Reagent
Chemical Name or Material Methyl iso-Butyl Ketone
Show More Show Less
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. Harmful by inhalation. Irritating to eyes, respiratory system and skin. May cause central nervous system effects. Repeated exposure may cause skin dryness or cracking. Aspiration hazard if swallowed - can enter lungs and cause damage. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. If peroxide formation is suspected, do not open or move container. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes.

NFPA

Health:2
Flammability:3
Instability:1
DOTInformation DOT Class 3, : Flammable Liquid