missing translation for 'onlineSavingsMsg'
Learn More

Methyl Indole-6-carboxylate 98.0+%, TCI America™

Catalog No. p-7132988
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M20421G 1 g
M20425G 5 g
M204225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M20421G Supplier TCI America Supplier No. M20421G
Only null left

Chemical Identifiers

CAS 50820-65-0
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.187
MDL Number MFCD00211063
InChI Key AYYOZKHMSABVRP-UHFFFAOYSA-N
Synonym methyl indole-6-carboxylate, indole-6-carboxylic acid methyl ester, methylindole-6-carboxylate, 1h-indole-6-carboxylic acid, methyl ester, 1h-indole-6-carboxylic acid methyl ester, indole-7-carboxylate, 6-carbomethoxyindole, indole-6-carboxylate, pubchem1696, 6-methoxycarbonylindole
PubChem CID 639844
IUPAC Name methyl 1H-indole-6-carboxylate
SMILES COC(=O)C1=CC2=C(C=C1)C=CN2

Specifications

Melting Point 77°C
Color White-Yellow
Quantity 1 g
Formula Weight 175.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl Indole-6-carboxylate