missing translation for 'onlineSavingsMsg'
Learn More

Methyl Indole-5-carboxylate 98.0+%, TCI America™

Catalog No. I05455G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I05455G 5 g
I05451G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I05455G Supplier TCI America Supplier No. I05455G

Chemical Identifiers

CAS 1011-65-0
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.187
MDL Number MFCD00153023
InChI Key DRYBMFJLYYEOBZ-UHFFFAOYSA-N
Synonym methyl indole-5-carboxylate, indole-5-carboxylic acid methyl ester, 1h-indole-5-carboxylic acid, methyl ester, methylindole-5-carboxylate, 1h-indole-5-carboxylic acid methyl ester, indole5carboxylicacidmethylester, indole-5-carboxylicacidmethylester, indole-5-carboxylate, pubchem7248, 5-methoxycarbonylindole
PubChem CID 2737635
IUPAC Name methyl 1H-indole-5-carboxylate
SMILES COC(=O)C1=CC2=C(C=C1)NC=C2

Specifications

Melting Point 126°C
Quantity 5 g
Formula Weight 175.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl Indole-5-carboxylate