missing translation for 'onlineSavingsMsg'
Learn More

Methyl indole-5-carboxylate, 97%

Catalog No. aal1622806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1622806 5 g
AAL1622803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1622806 Supplier Thermo Scientific Chemicals Supplier No. L1622806
Only null left

CAS: 1011-65-0 | C10H9NO2 | 175.187 g/mol

Reactant for biosynthesis of inhibitors of protein kinases, reactant in metal-free Friedel-Crafts alkylation, reactant in preparation of diphenylsulfonium ylides from Martin?s sulfurane, reactant in cross dehydrogenative coupling reactions, reactant in synthesis of indirubin derivatives, reactant in preparation of aminoindolylacetates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant for biosynthesis of inhibitors of protein kinases, reactant in metal-free Friedel-Crafts alkylation, reactant in preparation of diphenylsulfonium ylides from Martin′s sulfurane, reactant in cross dehydrogenative coupling reactions, reactant in synthesis of indirubin derivatives, reactant in preparation of aminoindolylacetates.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 1011-65-0
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.187
MDL Number MFCD00153023
InChI Key DRYBMFJLYYEOBZ-UHFFFAOYSA-N
Synonym methyl indole-5-carboxylate, indole-5-carboxylic acid methyl ester, 1h-indole-5-carboxylic acid, methyl ester, methylindole-5-carboxylate, 1h-indole-5-carboxylic acid methyl ester, indole5carboxylicacidmethylester, indole-5-carboxylicacidmethylester, indole-5-carboxylate, pubchem7248, 5-methoxycarbonylindole
PubChem CID 2737635
IUPAC Name methyl 1H-indole-5-carboxylate
SMILES COC(=O)C1=CC2=C(C=C1)NC=C2

Specifications

Melting Point 123°C to 126°C
Quantity 5 g
Beilstein 474256
Solubility Information Insoluble in water.
Formula Weight 175.19
Percent Purity ≥98%
Chemical Name or Material Methyl indole-5-carboxylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only