missing translation for 'onlineSavingsMsg'
Learn More

Methyl Indole-4-carboxylate 98.0+%, TCI America™

Catalog No. M17731G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M17731G 1 g
M17735G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M17731G Supplier TCI America Supplier No. M17731G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 39830-66-5
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.187
MDL Number MFCD00191222
InChI Key WEAXQUBYRSEBJD-UHFFFAOYSA-N
Synonym methyl indole-4-carboxylate, indole-4-carboxylic acid methyl ester, 1h-indole-4-carboxylic acid methyl ester, methyl 4-indolecarboxylate, 4-indole-carboxylic acid methyl ester, 1h-indole-4-carboxylic acid, methyl ester, 4-methoxycarbonylindole, 4-methoxycarbonyl-1h-indole, indole-4-carboxylate, pubchem1705
PubChem CID 2733668
IUPAC Name methyl 1H-indole-4-carboxylate
SMILES COC(=O)C1=C2C=CNC2=CC=C1

Specifications

Melting Point 71°C
Quantity 1 g
Formula Weight 175.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl Indole-4-carboxylate