missing translation for 'onlineSavingsMsg'
Learn More

Methyl Indole-3-carboxylate 98.0+%, TCI America™

Catalog No. p-7131437
Change view
Click to view available options
Quantity:
25 g
500 g
Catalog No. Quantity
I049125G 25 g
I0491500G 500 g
2 options

Catalog No. I049125G

Supplier: TCI America I049125G

Only null left

Chemical Identifiers

CAS 942-24-5
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.187
MDL Number MFCD00189407
InChI Key QXAUTQFAWKKNLM-UHFFFAOYSA-N
Synonym methyl indole-3-carboxylate, indole-3-carboxylic acid methyl ester, methyl 3-indolecarboxylate, 1h-indole-3-carboxylic acid methyl ester, 1h-indole-3-carboxylic acid, methyl ester, 3-carbomethoxyindole, 3-methoxycarbonylindole, methylindole-3-carboxylate, indole-3-carboxylic acid, methyl ester, methyl-3-indolcarboxylate
PubChem CID 589098
ChEBI CHEBI:65019
IUPAC Name methyl 1H-indole-3-carboxylate
SMILES COC(=O)C1=CNC2=CC=CC=C21

Specifications

Melting Point 149°C
Quantity 25 g
Formula Weight 175.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl Indole-3-carboxylate