missing translation for 'onlineSavingsMsg'
Learn More

Methyl indole-3-carboxylate, 99%

Catalog No. AAL1451614
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1451614 25 g
AAL1451622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1451614 Supplier Thermo Scientific Chemicals Supplier No. L1451614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 942-24-5 | C10H9NO2 | 175.187 g/mol

Reactant used for preparation of nitric oxide synthesis (nNOS) inhibitors, protein kinase c alpha (PKCα) inhibitors, inhibitors of the C-terminal domain of RNA polymerase II as antitumor agents, kinase insert domain receptor (KDR) inhibitors, organocatalysts for the anti-Mannich reaction, very late antigen-4 (VLA-4) antagonists, inhibitors of Human 5-Lipoxygenase, serotonin 5-HT4 receptor antagonists and hyaluronidase inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant used for preparation of nitric oxide synthesis (nNOS) inhibitors, protein kinase c alpha (PKCα) inhibitors, inhibitors of the C-terminal domain of RNA polymerase II as antitumor agents, kinase insert domain receptor (KDR) inhibitors, organocatalysts for the anti-Mannich reaction, very late antigen-4 (VLA-4) antagonists, inhibitors of Human 5-Lipoxygenase, serotonin 5-HT4 receptor antagonists and hyaluronidase inhibitors.

Solubility
Slightly soluble methanol and dimethyl sulfoxide. Insoluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 942-24-5
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.187
MDL Number MFCD00189407
InChI Key QXAUTQFAWKKNLM-UHFFFAOYSA-N
Synonym methyl indole-3-carboxylate, indole-3-carboxylic acid methyl ester, methyl 3-indolecarboxylate, 1h-indole-3-carboxylic acid methyl ester, 1h-indole-3-carboxylic acid, methyl ester, 3-carbomethoxyindole, 3-methoxycarbonylindole, methylindole-3-carboxylate, indole-3-carboxylic acid, methyl ester, methyl-3-indolcarboxylate
PubChem CID 589098
ChEBI CHEBI:65019
IUPAC Name methyl 1H-indole-3-carboxylate
SMILES COC(=O)C1=CNC2=CC=CC=C21

Specifications

Melting Point 149°C to 152°C
Quantity 25 g
Beilstein 142023
Solubility Information Slightly soluble methanol and dimethyl sulfoxide. Insoluble in water.
Formula Weight 175.19
Percent Purity 99%
Chemical Name or Material Methyl indole-3-carboxylate
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only