missing translation for 'onlineSavingsMsg'
Learn More

Methyl Indole-2-carboxylate 98.0+%, TCI America™

Catalog No. p-7132838
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
M17495G 5 g
M174925G 25 g
2 options

Catalog No. M17495G

Supplier: TCI America M17495G

Only null left

Chemical Identifiers

CAS 1202-04-6
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.19
MDL Number MFCD00460779
InChI Key NQPIEWBAWBFGOB-UHFFFAOYSA-N
Synonym methyl indole-2-carboxylate, indole-2-carboxylic acid methyl ester, methyl 2-indolecarboxylate, 1h-indole-2-carboxylic acid methyl ester, 1h-indole-2-carboxylic acid, methyl ester, ethyl-indole-2-caboxylate, pubchem7284, 2-methoxycarbonylindole, methylindole-2-carboxylate, buttpark 99\04-61
PubChem CID 70992
IUPAC Name methyl 1H-indole-2-carboxylate
SMILES COC(=O)C1=CC2=CC=CC=C2N1

Specifications

Melting Point 153°C
Color White-Yellow
Quantity 5 g
Formula Weight 175.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl Indole-2-carboxylate