missing translation for 'onlineSavingsMsg'
Learn More

Methyl indole-2-carboxylate, 97%

Catalog No. p-7057186
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6660903 1 g
AAH6660906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6660903 Supplier Thermo Scientific Chemicals Supplier No. H6660903

CAS: 1202-04-6 | C10H9NO2 | 175.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1202-04-6
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.19
MDL Number MFCD00460779
InChI Key NQPIEWBAWBFGOB-UHFFFAOYSA-N
Synonym methyl indole-2-carboxylate, indole-2-carboxylic acid methyl ester, methyl 2-indolecarboxylate, 1h-indole-2-carboxylic acid methyl ester, 1h-indole-2-carboxylic acid, methyl ester, ethyl-indole-2-caboxylate, pubchem7284, 2-methoxycarbonylindole, methylindole-2-carboxylate, buttpark 99\04-61
PubChem CID 70992
IUPAC Name methyl 1H-indole-2-carboxylate
SMILES COC(=O)C1=CC2=CC=CC=C2N1

Specifications

Quantity 1 g
Beilstein 142184
Formula Weight 175.19
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Methyl indole-2-carboxylate
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 214-861-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only