missing translation for 'onlineSavingsMsg'
Learn More

Methyl Hydroxypivalate 98.0+%, TCI America™

Catalog No. H090625G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H090625G 25 g
H0906500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H090625G Supplier TCI America Supplier No. H090625G

Chemical Identifiers

CAS 14002-80-3
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.16
MDL Number MFCD00009707
InChI Key KJRFTNVYOAGTHK-UHFFFAOYSA-N
Synonym hydroxypivalic acid methyl ester, methyl 3-hydroxypivalate, methyl hydroxypivalate, methyl 2,2-dimethyl-3-hydroxypropionate, hydroxypivalicacidmethylester, propanoic acid, 3-hydroxy-2,2-dimethyl-, methyl ester, 2,2-dimethyl-3-hydroxypropionic acid methyl ester, 3-hydroxy-2,2-dimethyl-propionic acid methyl ester, methyl 2,2-dimethyl-3-hydroxypropanoate, methyl hydroxylpivalate
PubChem CID 84152
IUPAC Name methyl 3-hydroxy-2,2-dimethylpropanoate
SMILES COC(=O)C(C)(C)CO

Specifications

Melting Point -16°C
Color Colorless
Boiling Point 178°C
Quantity 25 g
Formula Weight 132.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Hydroxypivalate
EINECSNumber (2)-1352
TSCA No