missing translation for 'onlineSavingsMsg'
Learn More

Methyl hydrogen suberate, 98%

CAS: 3946-32-5 | C9H15O4 | 187.22 g/mol

$111.58 - $1115.67

Chemical Identifiers

CAS 3946-32-5
Molecular Formula C9H15O4
Molecular Weight (g/mol) 187.22
MDL Number MFCD00004427
InChI Key KOVPXZDUVJGGFU-UHFFFAOYSA-M
Synonym suberic acid monomethyl ester, monomethyl suberate, methyl hydrogen suberate, subericacidmonomethylester, octanedioic acid monomethyl ester, mono-methyl suberate, acmc-1ckw1, ksc492m6f, 7-methoxycarbonylheptanoic acid, octanedioic acid 1-methyl ester
PubChem CID 554191
IUPAC Name 8-methoxy-8-oxooctanoic acid
SMILES COC(=O)CCCCCCC([O-])=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH3032503
View Documents
Thermo Scientific Chemicals
H3032503
1 g
Each for $111.58
Only null left
 
AAH3032506
View Documents
Thermo Scientific Chemicals
H3032506
5 g
Each for $330.11
Only null left
 
AAH3032514
View Documents
Thermo Scientific Chemicals
H3032514
25 g
Each for $1,115.67
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3946-32-5
187.22
KOVPXZDUVJGGFU-UHFFFAOYSA-M
554191
COC(=O)CCCCCCC([O-])=O
C9H15O4
MFCD00004427
suberic acid monomethyl ester, monomethyl suberate, methyl hydrogen suberate, subericacidmonomethylester, octanedioic acid monomethyl ester, mono-methyl suberate, acmc-1ckw1, ksc492m6f, 7-methoxycarbonylheptanoic acid, octanedioic acid 1-methyl ester
8-methoxy-8-oxooctanoic acid

Specifications

3946-32-5
1.04
C9H15O4
MFCD00004427
suberic acid monomethyl ester, monomethyl suberate, methyl hydrogen suberate, subericacidmonomethylester, octanedioic acid monomethyl ester, mono-methyl suberate, acmc-1ckw1, ksc492m6f, 7-methoxycarbonylheptanoic acid, octanedioic acid 1-methyl ester
COC(=O)CCCCCCC([O-])=O
187.22
188.22
Methyl hydrogen suberate
17°C to 19°C
185°C to 186°C (18 mmHg)
CH3O2C(CH2)6CO2H
1 g
KOVPXZDUVJGGFU-UHFFFAOYSA-M
8-methoxy-8-oxooctanoic acid
554191
98%
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only