missing translation for 'onlineSavingsMsg'
Learn More

Methyl hydrogen malonate, 96%

Catalog No. p-7030217
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2777206 5 g
AAH2777214 25 g
AAH2777222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH2777206 Supplier Thermo Scientific Chemicals Supplier No. H2777206

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 16695-14-0 | C4H6O4 | 118.088 g/mol

Lactonization of olefins with monomethyl ester of malonic acid and ceric ammonium nitrate was carried out in acetic acid and in acetonitrile under simple mechanical stirring or ultrasound irradiation. Lactonization of olefins with CAN and monomethyl ester of malonic acid under ultrasound irradiation shows an acceleration with respect to the analogous mechanically stirred reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lactonization of olefins with monomethyl ester of malonic acid and ceric ammonium nitrate was carried out in acetic acid and in acetonitrile under simple mechanical stirring or ultrasound irradiation. Lactonization of olefins with CAN and monomethyl ester of malonic acid under ultrasound irradiation shows an acceleration with respect to the analogous mechanically stirred reaction.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 16695-14-0
Molecular Formula C4H6O4
Molecular Weight (g/mol) 118.088
MDL Number MFCD00667796
InChI Key PBVZQAXFSQKDKK-UHFFFAOYSA-N
Synonym methyl hydrogen malonate, malonic acid monomethyl ester, 3-methoxy-3-oxo-propanoic acid, 1-methyl malonate, hydrogen methyl malonate
PubChem CID 538366
IUPAC Name 3-methoxy-3-oxopropanoic acid
SMILES COC(=O)CC(=O)O

Specifications

Density 1.128
Refractive Index 1.4295
Quantity 5 g
Formula Weight 118.09
Percent Purity 96%
Chemical Name or Material Methyl hydrogen malonate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only