missing translation for 'onlineSavingsMsg'
Learn More

Methyl Heptanoate 97.0+%, TCI America™

Catalog No. H003225ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H003225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H003225ML Supplier TCI America Supplier No. H003225ML
Only null left

Specifications

CAS 106-73-0
Color Yellow
Boiling Point 172°C
Molecular Formula C8H16O2
Quantity 25 mL
MDL Number MFCD00009537
UN Number 3272
Synonym methyl enanthate, heptanoic acid, methyl ester, methyl oenanthylate, methyl heptoate, methyl heptylate, heptanoic acid methyl ester, methyl n-heptylate, enanthic acid methyl ester, methyl heptylate natural, methyl n-heptanoate
InChI Key XNCNNDVCAUWAIT-UHFFFAOYSA-N
SMILES CCCCCCC(=O)OC
IUPAC Name methyl heptanoate
Molecular Weight (g/mol) 144.214
PubChem CID 7826
Formula Weight 144.21
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Heptanoate
Show More Show Less
EINECSNumber (2)-0782
RTECSNumber MJ2297500
TSCA Yes