missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Methyl Green, zinc chloride salt, pure, certified

Catalog No. p-4229039
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC414380250 25 g Glass bottle
AC414381000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC414380250 Supplier Thermo Scientific Chemicals Supplier No. 414380250
Only null left

CAS: 7114-03-6 | C27H35BrClN3·xZnCl2 | 653.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7114-03-6
Molecular Formula C27H35BrClN3·xZnCl2
Molecular Weight (g/mol) 653.23

Specifications

Melting Point 233.0°C
Color Brown-Red or Green
Physical Form Crystalline Powder
Quantity 25 g
Merck Index 14,6082
Synonym C.I. 42590, Ethyl Green, zinc chloride salt
Solubility Information Solubility in water: 70g/L (20°C)
InChI Key ZKOGXCSOLMZRAZ-UHFFFAOYSA-J
SMILES CC[N+](C)(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-].[Cl-].[Cl-].[Zn+2].[Br-]
IUPAC Name zinc;[4-[[4-(dimethylamino)phenyl]-(4-dimethylazaniumylidenecyclohexa-2,5-dien-1-ylidene)methyl]phenyl]-ethyl-dimethylazanium;bromide;trichloride
PubChem CID 131675247
Formula Weight 653.23
Grade Pure
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Methyl Green, zinc chloride salt
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 230-415-4

RUO – Research Use Only