missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Methyl Green, zinc chloride

Catalog No. AAJ6150914
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6150914 25 g
AAJ6150922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6150914 Supplier Thermo Scientific Chemicals Supplier No. J6150914
Only null left

CAS: 7114-03-6 | C27H35Cl4N3Zn | 608.78 g/mol

Methyl Green is a DNA stain used as a tracking dye in acid buffers for electrophoresis. It helps differentiate bacteria and is also a tissue stain for nucleoli and mitochondria in tissues. It is used with Bismarck Brown frequently.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl Green is a DNA stain used as a tracking dye in acid buffers for electrophoresis. It helps differentiate bacteria and is also a tissue stain for nucleoli and mitochondria in tissues. It is used with Bismarck Brown frequently.

Solubility
Soluble in water, ethanol or methanol

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 7114-03-6
Molecular Formula C27H35Cl4N3Zn
Molecular Weight (g/mol) 608.78
MDL Number MFCD00151094
InChI Key ZKOGXCSOLMZRAZ-UHFFFAOYSA-J
Synonym C.I. 42590
PubChem CID 131675247
IUPAC Name zinc;[4-[[4-(dimethylamino)phenyl]-(4-dimethylazaniumylidenecyclohexa-2,5-dien-1-ylidene)methyl]phenyl]-ethyl-dimethylazanium;bromide;trichloride
SMILES CC[N+](C)(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-].[Cl-].[Cl-].[Zn+2].[Br-]

Specifications

Melting Point >300°C
Beilstein 3914106
Solubility Information Soluble in water,ethanol or methanol
Formula Weight 653.24
Color Green
Quantity 25 g
Physical Form Powder
Chemical Name or Material Methyl Green, zinc chloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 230-415-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only