missing translation for 'onlineSavingsMsg'
Learn More

Methyl Glycolate 98.0+%, TCI America™

Catalog No. G019925G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G019925G 25 g
G0199100G 100 g
G0199500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. G019925G Supplier TCI America Supplier No. G019925G

Chemical Identifiers

CAS 96-35-5
Molecular Formula C3H6O3
Molecular Weight (g/mol) 90.078
MDL Number MFCD00004667
InChI Key GSJFXBNYJCXDGI-UHFFFAOYSA-N
Synonym methyl glycolate, methyl hydroxyacetate, acetic acid, hydroxy-, methyl ester, glycolic acid, methyl ester, acetic acid, 2-hydroxy-, methyl ester, glycolic acid methyl ester, unii-7g7r729ej0, hydroxyacetic acid methyl ester, glycolic acid methyl, methyl2-hydroxyethanoate
PubChem CID 66774
IUPAC Name methyl 2-hydroxyacetate
SMILES COC(=O)CO

Specifications

Color Colorless
Boiling Point 151°C
Quantity 25 g
Formula Weight 90.08
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Glycolate
EINECSNumber (2)-3774
TSCA Yes