missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Gallate 98.0+%, TCI America™
$46.20 - $425.46
Chemical Identifiers
| CAS | 99-24-1 |
|---|---|
| Molecular Formula | C8H8O5 |
| Molecular Weight (g/mol) | 184.147 |
| MDL Number | MFCD00002194 |
| InChI Key | FBSFWRHWHYMIOG-UHFFFAOYSA-N |
| Synonym | methyl gallate, gallic acid methyl ester, methylgallate, benzoic acid, 3,4,5-trihydroxy-, methyl ester, gallic acid, methyl ester, methyl galloate, unii-623d3xg80c, ccris 5567, 3,4,5-trihydroxybenzoic acid methyl ester, chembl65675 |
| PubChem CID | 7428 |
| IUPAC Name | methyl 3,4,5-trihydroxybenzoate |
| SMILES | COC(=O)C1=CC(=C(C(=C1)O)O)O |
Chemical Identifiers
| 99-24-1 | |
| 184.147 | |
| FBSFWRHWHYMIOG-UHFFFAOYSA-N | |
| 7428 | |
| COC(=O)C1=CC(=C(C(=C1)O)O)O |
| C8H8O5 | |
| MFCD00002194 | |
| methyl gallate, gallic acid methyl ester, methylgallate, benzoic acid, 3,4,5-trihydroxy-, methyl ester, gallic acid, methyl ester, methyl galloate, unii-623d3xg80c, ccris 5567, 3,4,5-trihydroxybenzoic acid methyl ester, chembl65675 | |
| methyl 3,4,5-trihydroxybenzoate |
Specifications
| 99-24-1 | |
| White-Yellow | |
| MFCD00002194 | |
| methyl gallate, gallic acid methyl ester, methylgallate, benzoic acid, 3,4,5-trihydroxy-, methyl ester, gallic acid, methyl ester, methyl galloate, unii-623d3xg80c, ccris 5567, 3,4,5-trihydroxybenzoic acid methyl ester, chembl65675 | |
| COC(=O)C1=CC(=C(C(=C1)O)O)O | |
| 184.147 | |
| 184.15 | |
| Crystalline Powder |
| 200°C | |
| C8H8O5 | |
| 25 g | |
| FBSFWRHWHYMIOG-UHFFFAOYSA-N | |
| methyl 3,4,5-trihydroxybenzoate | |
| 7428 | |
| ≥98.0% (GC,T) | |
| Methyl Gallate |
Safety and Handling
EINECSNumber : (3)-1583
RTECSNumber : LW8000000
TSCA : Yes