missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Formate 95.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 32°C |
| Quantity | 25 mL |
| UN Number | 1243 |
| Formula Weight | 60.05 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Methyl Formate |
Safety and Handling
| EINECSNumber | (2)-0677 |
| RTECSNumber | LQ8925000 |
| TSCA | Yes |