missing translation for 'onlineSavingsMsg'
Learn More

Methyl Formate 95.0+%, TCI America™

Catalog No. F005725ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F005725ML 25 mL
F0057500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F005725ML Supplier TCI America Supplier No. F005725ML

Chemical Identifiers

CAS 107-31-3
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00003291
InChI Key TZIHFWKZFHZASV-UHFFFAOYSA-N
Synonym formic acid, methyl ester, methyl methanoate, methylformiat, formic acid methyl ester, formiate de methyle, methylformiaat, mravencan methylnaty, methylformate, caswell no. 570, hcooch3
PubChem CID 7865
ChEBI CHEBI:77699
IUPAC Name methyl formate
SMILES COC=O

Specifications

Color Colorless
Boiling Point 32°C
Quantity 25 mL
UN Number 1243
Formula Weight 60.05
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Formate
EINECSNumber (2)-0677
RTECSNumber LQ8925000
TSCA Yes