missing translation for 'onlineSavingsMsg'
Learn More

Methyl formate, 98%, for spectroscopy

CAS: 107-31-3 | C2H4O2 | 60.05 g/mol

$236.57 - $236.57

Chemical Identifiers

CAS 107-31-3
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00003291
InChI Key TZIHFWKZFHZASV-UHFFFAOYSA-N
Synonym formic acid, methyl ester, methyl methanoate, methylformiat, formic acid methyl ester, formiate de methyle, methylformiaat, mravencan methylnaty, methylformate, caswell no. 570, hcooch3
PubChem CID 7865
ChEBI CHEBI:77699
IUPAC Name methyl formate
SMILES COC=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC414345000
SDS View Documents
Thermo Scientific Chemicals
414345000
500 mL Glass bottle
Each for $236.57
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

for UV/VIS spectroscopy
Specifications

Chemical Identifiers

107-31-3
60.05
TZIHFWKZFHZASV-UHFFFAOYSA-N
7865
methyl formate
C2H4O2
MFCD00003291
formic acid, methyl ester, methyl methanoate, methylformiat, formic acid methyl ester, formiate de methyle, methylformiaat, mravencan methylnaty, methylformate, caswell no. 570, hcooch3
CHEBI:77699
COC=O

Specifications

107-31-3
0.009 max. at 320nm, 0.032 max. at 300nm, 0.051 max. at 270nm, 1 max. at 260nm
0.9680g/mL
−32°C
97.5% min. (GC)
C2H4O2
HCO2CH3
500 mL
0.0005% max.
0.968
formic acid, methyl ester, methyl methanoate, methylformiat, formic acid methyl ester, formiate de methyle, methylformiaat, mravencan methylnaty, methylformate, caswell no. 570, hcooch3
TZIHFWKZFHZASV-UHFFFAOYSA-N
methyl formate
7865
CHEBI:77699
98%
Liquid
0.0008 meq/g max.
-100.0°C
31.0°C to 33.0°C
Authentic
Glass bottle
1.3420 to 1.3440
MFCD00003291
02, 18
05,446
15, 6148
Solubility in water: 300g/L (20°C). Other solubilities: soluble in almost all conventional organic solvent,miscible with alcohol,soluble in ether
COC=O
60.05
0.35 mPa.s (20°C)
60.05
Spectroscopy
Methyl formate, 98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful if inhaled.
Extremely flammable liquid and vapor.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
If exposed or you

GHS Signal Word: Danger

EINECSNumber : 203-481-7

RTECSNumber : LQ8925000

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only