missing translation for 'onlineSavingsMsg'
Learn More

Methyl formate, 98%, for spectroscopy

Catalog No. AC414340050
Change view
Click to view available options
Quantity:
5 mL
500 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC414340050 5 mL Glass bottle
AC414345000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC414340050 Supplier Thermo Scientific Chemicals Supplier No. 414340050

CAS: 107-31-3 | C2H4O2 | 60.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

for UV/VIS spectroscopy

Chemical Identifiers

CAS 107-31-3
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00003291
InChI Key TZIHFWKZFHZASV-UHFFFAOYSA-N
Synonym formic acid, methyl ester, methyl methanoate, methylformiat, formic acid methyl ester, formiate de methyle, methylformiaat, mravencan methylnaty, methylformate, caswell no. 570, hcooch3
PubChem CID 7865
ChEBI CHEBI:77699
IUPAC Name methyl formate
SMILES COC=O

Specifications

Acidity 0.0008 meq/g max.
Absorbance 0.009 max. at 320nm, 0.032 max. at 300nm, 0.051 max. at 270nm, 1 max. at 260nm
Melting Point -100.0°C
Density 0.9680g/mL
Boiling Point 31.0°C to 33.0°C
Flash Point −32°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.3420 to 1.3440
Linear Formula HCO2CH3
Quantity 5 mL
Residue after Evaporation 0.0005% max.
Beilstein 02, 18
Fieser 05,446
Specific Gravity 0.968
Merck Index 15, 6148
Solubility Information Solubility in water: 300g/L (20°C). Other solubilities: soluble in almost all conventional organic solvent,miscible with alcohol,soluble in ether
Viscosity 0.35 mPa.s (20°C)
Formula Weight 60.05
Percent Purity 98%
Grade Spectroscopy
Physical Form Liquid
Chemical Name or Material Methyl formate, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful if inhaled.
Extremely flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
If exposed or you
EINECSNumber 203-481-7
RTECSNumber LQ8925000
TSCA TSCA

RUO – Research Use Only