missing translation for 'onlineSavingsMsg'
Learn More

Methyl Ethyl Ketone (ACS), Fisher Chemical™

Catalog No. p-216191
Change view
Click to view available options
Quantity:
1 L
4 L
200 L
Packaging:
Glass Bottle
Steel Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M2091 1 L Glass Bottle
M2094 4 L Glass Bottle
M209200 200 L Steel Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M2091 Supplier Thermo Fisher Scientific Formerly Fairlawn Supplier No. M209 1
Only null left

Chemical Identifiers

CAS 78-93-3
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00011648
InChI Key ZWEHNKRNPOVVGH-UHFFFAOYSA-N
Synonym 2-butanone, methyl ethyl ketone, butanone, ethyl methyl ketone, methylethyl ketone, meetco, methyl acetone, methylethylketone, 3-butanone, ethylmethylketon
PubChem CID 6569
ChEBI CHEBI:28398
IUPAC Name butan-2-one
SMILES CCC(C)=O

Specifications

Melting Point -87°C
Color Colorless
Density 0.806g/cm³
Quantity 1 L
Identification Pass Test
Packaging Glass Bottle
Physical Form Liquid
Residue after Evaporation 0.0025% max.
Assay Percent Range ≥99 %
Titratable Acid 0.0005mEq/g max.
Vapor Pressure 105mbar at 20°C
Viscosity 0.42 mPa/s at 15°C
Water 0.1% max.
Boiling Point 80°C
Formula Weight 72.11
Percent Purity ≥99%
Grade ACS
Chemical Name or Material Methyl Ethyl Ketone
Show More Show Less
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. Vapors may cause drowsiness and dizziness. May cause irritation of respiratory tract. Repeated exposure may cause skin dryness or cracking. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:1
Flammability:3
Instability:0
DOTInformation DOT Class 3, : Flammable Liquid