missing translation for 'onlineSavingsMsg'
Learn More

Methyl Ethyl Ketone, ACS Reagent (2-Butanone), Reagents

Catalog No. 01337889
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337889 10 L
01337888 1 L
01337887 20 L
01337886 4 L
01337885 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 01337889 Supplier Labchem Inc Supplier No. C125050010E2

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Methyl Ethyl Ketone, ACS Reagent (2-Butanone), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 78-93-3
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
InChI Key ZWEHNKRNPOVVGH-UHFFFAOYSA-N
Synonym 2-Butanone
IUPAC Name butan-2-one
SMILES CCC(C)=O

Specifications

Melting Point -86.4°C
Color Colorless
Quantity 10 L
Packaging HDPE Winpak
Physical Form Liquid
Specific Gravity 0.805
Boiling Point 80°C
Grade ACS
Chemical Name or Material Methyl Ethyl Ketone, ACS Reagent (2-Butanone)