missing translation for 'onlineSavingsMsg'
Learn More

Methyl DL-Pyroglutamate 98.0+%, TCI America™

Catalog No. p-7133387
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M27201G 1 g
M27205G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M27201G Supplier TCI America Supplier No. M27201G

Chemical Identifiers

CAS 54571-66-3
Molecular Formula C6H9NO3
Molecular Weight (g/mol) 143.14
MDL Number MFCD03844651
InChI Key HQGPKMSGXAUKHT-UHFFFAOYNA-N
Synonym methyl dl-pyroglutamate, 5-oxo-pyrrolidine-2-carboxylic acid methyl ester, methyl 5-oxo-2-pyrrolidinecarboxylate, 5-oxo-2-pyrrolidinecarboxylic acid methyl ester, methyl 5-oxo-dl-prolinate, methyl 5-oxo-2-pyrrolidinecarboxylate #, proline, 5-oxo-, methylester, 5-oxo-dl-proline methyl ester, methyl 2-pyrolidone-5-carboxylate, dl-pyroglutamic acid methyl ester
PubChem CID 500249
IUPAC Name methyl 5-oxopyrrolidine-2-carboxylate
SMILES COC(=O)C1CCC(=O)N1

Specifications

Boiling Point 135°C
Quantity 1 g
Formula Weight 143.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl DL-Pyroglutamate