missing translation for 'onlineSavingsMsg'
Learn More

Methyl DL-2-bromopropionate, 99%, Thermo Scientific™

Chemical Identifiers

CAS 5445-17-0
Molecular Formula C4H7BrO2
Molecular Weight (g/mol) 167
MDL Number MFCD00000143
InChI Key ACEONLNNWKIPTM-UHFFFAOYSA-N
Synonym methyl 2-bromopropionate, propanoic acid, 2-bromo-, methyl ester, 2-bromopropionic acid methyl ester, methyl dl-2-bromopropionate, methyl2-bromopropionate, methyl alpha-bromopropionate, methyl .alpha.-bromopropionate, methyl-2-bromopropanoate, methyl 2-bromo-propionate, propionic acid, 2-bromo-, methyl ester
PubChem CID 95576
IUPAC Name methyl 2-bromopropanoate
SMILES CC(C(=O)OC)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC173391000
SDS View Documents
Thermo Scientific Chemicals
173391000
100g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC173395000
SDS View Documents
Thermo Scientific Chemicals
173395000
500g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

5445-17-0
167
ACEONLNNWKIPTM-UHFFFAOYSA-N
95576
CC(C(=O)OC)Br
C4H7BrO2
MFCD00000143
methyl 2-bromopropionate, propanoic acid, 2-bromo-, methyl ester, 2-bromopropionic acid methyl ester, methyl dl-2-bromopropionate, methyl2-bromopropionate, methyl alpha-bromopropionate, methyl .alpha.-bromopropionate, methyl-2-bromopropanoate, methyl 2-bromo-propionate, propionic acid, 2-bromo-, methyl ester
methyl 2-bromopropanoate

Specifications

5445-17-0
143.0°C to 145.0°C (19.0mmHg)
Authentic
Glass bottle
1.4509 to 1.4530
MFCD00000143
02, 253
methyl 2-bromopropionate, propanoic acid, 2-bromo-, methyl ester, 2-bromopropionic acid methyl ester, methyl dl-2-bromopropionate, methyl2-bromopropionate, methyl alpha-bromopropionate, methyl .alpha.-bromopropionate, methyl-2-bromopropanoate, methyl 2-bromo-propionate, propionic acid, 2-bromo-, methyl ester
ACEONLNNWKIPTM-UHFFFAOYSA-N
methyl 2-bromopropanoate
95576
99%
1.4970g/mL
51°C
98.5% min. (GC)
C4H7BrO2
CH3CH(Br)CO2CH3
100g
1.497
Solubility in water: immiscible
CC(C(=O)OC)Br
167
167
Methyl DL-2-bromopropionate, 99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapour.
Causes severe skin burns and eye damage.
May cause respiratory irritation.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomi

GHS Signal Word: Danger

EINECSNumber : 226-642-3

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only