missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl DL-2-Aminobutyrate Hydrochloride 98.0+%, TCI America™
$182.68 - $656.14
Chemical Identifiers
| CAS | 7682-18-0 |
|---|---|
| Molecular Formula | C5H12ClNO2 |
| Molecular Weight (g/mol) | 153.606 |
| MDL Number | MFCD00058295 |
| InChI Key | AHAQQEGUPULIOZ-UHFFFAOYSA-N |
| Synonym | DL-2-Aminobutyric Acid Methyl Ester Hydrochloride |
| PubChem CID | 13283515 |
| IUPAC Name | methyl 2-aminobutanoate;hydrochloride |
| SMILES | CCC(C(=O)OC)N.Cl |
Chemical Identifiers
| 7682-18-0 | |
| 153.606 | |
| AHAQQEGUPULIOZ-UHFFFAOYSA-N | |
| 13283515 | |
| CCC(C(=O)OC)N.Cl |
| C5H12ClNO2 | |
| MFCD00058295 | |
| DL-2-Aminobutyric Acid Methyl Ester Hydrochloride | |
| methyl 2-aminobutanoate;hydrochloride |
Specifications
| 7682-18-0 | |
| White | |
| MFCD00058295 | |
| DL-2-Aminobutyric Acid Methyl Ester Hydrochloride | |
| CCC(C(=O)OC)N.Cl | |
| 153.606 | |
| 153.61 | |
| Crystalline Powder |
| 150°C | |
| C5H12ClNO2 | |
| 5 g | |
| AHAQQEGUPULIOZ-UHFFFAOYSA-N | |
| methyl 2-aminobutanoate;hydrochloride | |
| 13283515 | |
| ≥98.0% (N) | |
| Methyl DL-2-Aminobutyrate Hydrochloride |
Safety and Handling
TSCA : No