missing translation for 'onlineSavingsMsg'
Learn More

Methyl dimethoxyacetate, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 414180050

Quantity 5g

 View more versions of this product

Catalog No. AC414180050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

89-91-8
134.13
NZTCVGHPDWAALP-UHFFFAOYSA-N
66647
COC(C(=O)OC)OC
C5H10O4
MFCD00008484
methyl dimethoxyacetate, acetic acid, dimethoxy-, methyl ester, dimethoxyacetic acid methyl ester, glyoxylic acid methyl ester dimethyl acetal, acetic acid, 2,2-dimethoxy-, methyl ester, glyoxylic acid, methyl ester, 2-dimethyl acetal, zlchem 519, acmc-209r3k, ksc490c4f, methyl 2,2-dimethoxyethanoate
methyl 2,2-dimethoxyacetate

Specifications

Methyl dimethoxyacetate
89-91-8
160°C to 163°C
Authentic
Glass Bottle
1.4040 to 1.406
5g
1.09
NZTCVGHPDWAALP-UHFFFAOYSA-N
methyl 2,2-dimethoxyacetate
66647
98%
99%
1.0900g/mL
63°C
97.5% min. (GC)
C5H10O4
(CH3O)2CHCOOCH3
MFCD00008484
methyl dimethoxyacetate, acetic acid, dimethoxy-, methyl ester, dimethoxyacetic acid methyl ester, glyoxylic acid methyl ester dimethyl acetal, acetic acid, 2,2-dimethoxy-, methyl ester, glyoxylic acid, methyl ester, 2-dimethyl acetal, zlchem 519, acmc-209r3k, ksc490c4f, methyl 2,2-dimethoxyethanoate
COC(C(=O)OC)OC
134.13
134.13
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

MOISTURE SENSITIVE

EINECSNumber : 201-950-0

TSCA : TSCA

SDS
Documents

Documents

Promotions

Promotions