missing translation for 'onlineSavingsMsg'
Learn More

Methyl dimethoxyacetate, 96%

Catalog No. AAA1217109
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1217109 10 g
AAA1217118 50 g
AAA1217130 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1217109 Supplier Thermo Scientific Chemicals Supplier No. A1217109

CAS: 89-91-8 | C5H10O4 | 134.131 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 89-91-8
Molecular Formula C5H10O4
Molecular Weight (g/mol) 134.131
MDL Number MFCD00008484
InChI Key NZTCVGHPDWAALP-UHFFFAOYSA-N
Synonym methyl dimethoxyacetate, acetic acid, dimethoxy-, methyl ester, dimethoxyacetic acid methyl ester, glyoxylic acid methyl ester dimethyl acetal, acetic acid, 2,2-dimethoxy-, methyl ester, glyoxylic acid, methyl ester, 2-dimethyl acetal, zlchem 519, acmc-209r3k, ksc490c4f, methyl 2,2-dimethoxyethanoate
PubChem CID 66647
IUPAC Name methyl 2,2-dimethoxyacetate
SMILES COC(C(=O)OC)OC

Specifications

Density 1.098
Boiling Point 160°C to 163°C
Flash Point 63°C (145°F)
Refractive Index 1.405
Quantity 10 g
Beilstein 1757582
Sensitivity Moisture sensitive
Formula Weight 134.13
Percent Purity ≥97%
Chemical Name or Material Methyl dimethoxyacetate
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 201-950-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only