missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Dimethoxyacetate 95.0+%, TCI America™
$180.81 - $180.81
Chemical Identifiers
| CAS | 89-91-8 |
|---|---|
| Molecular Formula | C5H10O4 |
| Molecular Weight (g/mol) | 134.131 |
| MDL Number | MFCD00008484 |
| InChI Key | NZTCVGHPDWAALP-UHFFFAOYSA-N |
| Synonym | methyl dimethoxyacetate, acetic acid, dimethoxy-, methyl ester, dimethoxyacetic acid methyl ester, glyoxylic acid methyl ester dimethyl acetal, acetic acid, 2,2-dimethoxy-, methyl ester, glyoxylic acid, methyl ester, 2-dimethyl acetal, zlchem 519, acmc-209r3k, ksc490c4f, methyl 2,2-dimethoxyethanoate |
| PubChem CID | 66647 |
| IUPAC Name | methyl 2,2-dimethoxyacetate |
| SMILES | COC(C(=O)OC)OC |
Chemical Identifiers
| 89-91-8 | |
| 134.131 | |
| NZTCVGHPDWAALP-UHFFFAOYSA-N | |
| 66647 | |
| COC(C(=O)OC)OC |
| C5H10O4 | |
| MFCD00008484 | |
| methyl dimethoxyacetate, acetic acid, dimethoxy-, methyl ester, dimethoxyacetic acid methyl ester, glyoxylic acid methyl ester dimethyl acetal, acetic acid, 2,2-dimethoxy-, methyl ester, glyoxylic acid, methyl ester, 2-dimethyl acetal, zlchem 519, acmc-209r3k, ksc490c4f, methyl 2,2-dimethoxyethanoate | |
| methyl 2,2-dimethoxyacetate |
Specifications
| 89-91-8 | |
| 67°C | |
| MFCD00008484 | |
| methyl dimethoxyacetate, acetic acid, dimethoxy-, methyl ester, dimethoxyacetic acid methyl ester, glyoxylic acid methyl ester dimethyl acetal, acetic acid, 2,2-dimethoxy-, methyl ester, glyoxylic acid, methyl ester, 2-dimethyl acetal, zlchem 519, acmc-209r3k, ksc490c4f, methyl 2,2-dimethoxyethanoate | |
| COC(C(=O)OC)OC | |
| 134.131 | |
| 134.13 | |
| Liquid |
| Yellow | |
| C5H10O4 | |
| 25 mL | |
| NZTCVGHPDWAALP-UHFFFAOYSA-N | |
| methyl 2,2-dimethoxyacetate | |
| 66647 | |
| ≥95.0% (GC) | |
| Methyl Dimethoxyacetate |
Safety and Handling
TSCA : Yes