missing translation for 'onlineSavingsMsg'
Learn More

Methyl Diethylphosphonoacetate 96.0+%, TCI America™

Catalog No. d287325g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
D287325G 25 g
D28735G 5 g
2 options

Catalog No. D287325G

Supplier: TCI America D287325G

Only null left

Chemical Identifiers

CAS 1067-74-9
Molecular Formula C7H15O5P
Molecular Weight (g/mol) 210.166
MDL Number MFCD00009081
InChI Key CTSAXXHOGZNKJR-UHFFFAOYSA-N
Synonym methyl diethylphosphonoacetate, methyl 2-diethoxyphosphoryl acetate, acetic acid, diethoxyphosphinyl-, methyl ester, diethylmethylphosphonoacetate, methyl diethoxyphosphoryl acetate, diethyl carbomethoxymethylphosphonate, methyl 2-diethoxycarbonyl acetate, methyldiethylphosphonoacetate, diethyl methylphosphonoacetate, ksc489e0f
PubChem CID 66113
IUPAC Name methyl 2-diethoxyphosphorylacetate
SMILES CCOP(=O)(CC(=O)OC)OCC

Specifications

Color Colorless
Boiling Point 129°C
Quantity 25 g
Formula Weight 210.17
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Diethylphosphonoacetate