missing translation for 'onlineSavingsMsg'
Learn More

Methyl diethylphosphonoacetate, 97%

Catalog No. AC169980500
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC169980500 50 g Glass bottle
AC169980100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. AC169980500 missing translation for 'mfr' Thermo Scientific Chemicals missing translation for 'supplierNo' 169980500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1067-74-9 | C7H15O5P | 210.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For C-C Bond Formation (Olefination)

Chemical Identifiers

CAS 1067-74-9
Molecular Formula C7H15O5P
Molecular Weight (g/mol) 210.17
MDL Number MFCD00009081
InChI Key CTSAXXHOGZNKJR-UHFFFAOYSA-N
Synonym methyl diethylphosphonoacetate, methyl 2-diethoxyphosphoryl acetate, acetic acid, diethoxyphosphinyl-, methyl ester, diethylmethylphosphonoacetate, methyl diethoxyphosphoryl acetate, diethyl carbomethoxymethylphosphonate, methyl 2-diethoxycarbonyl acetate, methyldiethylphosphonoacetate, diethyl methylphosphonoacetate, ksc489e0f
PubChem CID 66113
IUPAC Name methyl 2-diethoxyphosphorylacetate
SMILES CCOP(=O)(CC(=O)OC)OCC

Specifications

Color Colorless to Yellow
Density 1.1400g/mL
Boiling Point 127.0°C to 131.0°C (9.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4320 to 1.4340
Linear Formula (C2H5O)2P(O)CH2CO2CH3
Quantity 50 g
Beilstein 04, II, 976
Specific Gravity 1.14
Formula Weight 210.17
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Methyl diethylphosphonoacetate, 98%
Show More Show Less
missing translation for 'healthHazard1' GHS Signal Word: Warning
missing translation for 'healthHazard2' GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
missing translation for 'healthHazard3' GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
EINECSNumber 213-938-2

RUO – Research Use Only