missing translation for 'onlineSavingsMsg'
Learn More

Methyl dichloroacetate, 99%

CAS: 116-54-1 | C3H4Cl2O2 | 142.97 g/mol

$63.19 - $63.19

Chemical Identifiers

CAS 116-54-1
Molecular Formula C3H4Cl2O2
Molecular Weight (g/mol) 142.97
MDL Number MFCD00000843
InChI Key HKMLRUAPIDAGIE-UHFFFAOYSA-N
Synonym methyl dichloroacetate, dichloroacetic acid methyl ester, acetic acid, dichloro-, methyl ester, ccris 7740, acetic acid, 2,2-dichloro-, methyl ester, methyldichloroethanoate, acmc-1brpm, dichloroacetic acid methyl, methyl2,2-dichloroacetate, wln: gygvo1
PubChem CID 8315
IUPAC Name methyl 2,2-dichloroacetate
SMILES COC(=O)C(Cl)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC148621000
SDS View Documents
Thermo Scientific Chemicals
148621000
100 mL Glass bottle
Each for $63.19
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

116-54-1
142.97
HKMLRUAPIDAGIE-UHFFFAOYSA-N
8315
COC(=O)C(Cl)Cl
C3H4Cl2O2
MFCD00000843
methyl dichloroacetate, dichloroacetic acid methyl ester, acetic acid, dichloro-, methyl ester, ccris 7740, acetic acid, 2,2-dichloro-, methyl ester, methyldichloroethanoate, acmc-1brpm, dichloroacetic acid methyl, methyl2,2-dichloroacetate, wln: gygvo1
methyl 2,2-dichloroacetate

Specifications

116-54-1
Colorless
141°C to 145°C
Authentic
Glass bottle
1.4411 to 1.4431
MFCD00000843
02, 203
methyl dichloroacetate, dichloroacetic acid methyl ester, acetic acid, dichloro-, methyl ester, ccris 7740, acetic acid, 2,2-dichloro-, methyl ester, methyldichloroethanoate, acmc-1brpm, dichloroacetic acid methyl, methyl2,2-dichloroacetate, wln: gygvo1
HKMLRUAPIDAGIE-UHFFFAOYSA-N
methyl 2,2-dichloroacetate
8315
99%
Methyl dichloroacetate, 99+%
-52°C
1.3810g/mL
80°C
98.5% min. (GC)
C3H4Cl2O2
Cl2CHCO2CH3
100 mL
1.381
Solubility in water: slightly soluble
COC(=O)C(Cl)Cl
142.97
142.97
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 204-146-8

RTECSNumber : AG6625000

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only