missing translation for 'onlineSavingsMsg'
Learn More

alpha-Methyl-D-mannopyranoside, 99+%

alpha-Methyl-D-mannopyranoside, 99+%, Quantity: 500g, Packaging: Glass bottle, Gray or White, Melting Point: 186.0 deg.C to 194.0 deg.C, Molecular Weight: 194.18, Percent Purity: 99+%, Appearance: (1% aq. soln.) clear, colorless, Assay Percent Range: 99% min. (ELSD) (HPLC) | CAS: 617-04-9 | C7H14O6 | 194.18 g/mol

$113.45 - $294.87

Chemical Identifiers

CAS 617-04-9
Molecular Formula C7H14O6
Molecular Weight (g/mol) 194.18
MDL Number MFCD00063262
InChI Key HOVAGTYPODGVJG-UHFFFAOYNA-N
Synonym methyl a-d-mannopyranoside, methyl alpha-d-mannopyranoside, methyl alpha-d-mannoside, alpha-methyl-d-mannoside, alpha-methyl mannopyranoside, 1-o-methyl-alpha-d-mannopyranoside, methyl-alpha-d-mannoside, alpha-d-methyl mannoside, o1-methyl-mannose, alpha-methyl-d---mannoside
PubChem CID 101798
ChEBI CHEBI:43943
IUPAC Name (2R,3S,4S,5S,6S)-2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol
SMILES COC1OC(CO)C(O)C(O)C1O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC229250250
SDS View Documents
Thermo Scientific Chemicals
229250250
25 g Glass bottle
Each for $113.45
Only null left
 
AC229251000
SDS View Documents
Thermo Scientific Chemicals
229251000
100 g Glass bottle
Each for $294.87
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

617-04-9
194.18
HOVAGTYPODGVJG-UHFFFAOYNA-N
101798
(2R,3S,4S,5S,6S)-2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol
C7H14O6
MFCD00063262
methyl a-d-mannopyranoside, methyl alpha-d-mannopyranoside, methyl alpha-d-mannoside, alpha-methyl-d-mannoside, alpha-methyl mannopyranoside, 1-o-methyl-alpha-d-mannopyranoside, methyl-alpha-d-mannoside, alpha-d-methyl mannoside, o1-methyl-mannose, alpha-methyl-d---mannoside
CHEBI:43943
COC1OC(CO)C(O)C(O)C1O

Specifications

617-04-9
100.0
Gray or White
99% min. (ELSD) (HPLC)
C7H14O6
25 g
+ 79.00
(10% in water) Clear colorless
COC1OC(CO)C(O)C(O)C1O
194.18
CHEBI:43943
99+%
α-Methyl-D-mannopyranoside
99.0
186.0°C to 194.0°C
Authentic
Glass bottle
MFCD00063262
+ 79.00 (20.00°C c=10,H2O)
methyl a-d-mannopyranoside, methyl alpha-d-mannopyranoside, methyl alpha-d-mannoside, alpha-methyl-d-mannoside, alpha-methyl mannopyranoside, 1-o-methyl-alpha-d-mannopyranoside, methyl-alpha-d-mannoside, alpha-d-methyl mannoside, o1-methyl-mannose, alpha-methyl-d---mannoside
HOVAGTYPODGVJG-UHFFFAOYNA-N
(2R,3S,4S,5S,6S)-2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol
101798
194.18
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 210-502-3

SDS
Documents

Documents

RUO – Research Use Only