missing translation for 'onlineSavingsMsg'
Learn More

alpha-Methyl-D-mannopyranoside, 99+%

Catalog No. AC229250250
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC229250250 25 g Glass bottle
AC229251000 100 g Glass bottle
AC229255000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC229250250 Supplier Thermo Scientific Chemicals Supplier No. 229250250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

alpha-Methyl-D-mannopyranoside, 99+%, Quantity: 500g, Packaging: Glass bottle, Gray or White, Melting Point: 186.0 deg.C to 194.0 deg.C, Molecular Weight: 194.18, Percent Purity: 99+%, Appearance: (1% aq. soln.) clear, colorless, Assay Percent Range: 99% min. (ELSD) (HPLC) | CAS: 617-04-9 | C7H14O6 | 194.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 617-04-9
Molecular Formula C7H14O6
Molecular Weight (g/mol) 194.18
MDL Number MFCD00063262
InChI Key HOVAGTYPODGVJG-UHFFFAOYNA-N
Synonym methyl a-d-mannopyranoside, methyl alpha-d-mannopyranoside, methyl alpha-d-mannoside, alpha-methyl-d-mannoside, alpha-methyl mannopyranoside, 1-o-methyl-alpha-d-mannopyranoside, methyl-alpha-d-mannoside, alpha-d-methyl mannoside, o1-methyl-mannose, alpha-methyl-d---mannoside
PubChem CID 101798
ChEBI CHEBI:43943
IUPAC Name (2R,3S,4S,5S,6S)-2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol
SMILES COC1OC(CO)C(O)C(O)C1O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 186.0°C to 194.0°C
Color Gray or White
Infrared Spectrum Authentic
Assay Percent Range 99% min. (ELSD) (HPLC)
Packaging Glass bottle
Quantity 25 g
Specific Rotation Condition + 79.00 (20.00°C c=10,H2O)
Specific Rotation + 79.00
Solubility Information (10% in water) Clear colorless
Formula Weight 194.18
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material α-Methyl-D-mannopyranoside
Show More Show Less
EINECSNumber 210-502-3

RUO – Research Use Only