missing translation for 'onlineSavingsMsg'
Learn More

Methyl α-D-mannopyranoside, ≥99%, Fisher BioReagents™

Catalog No. BP2530500
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
Packaging:
Amber Glass
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP2530500 500 g Amber Glass
BP253010 10 g Amber Glass
BP2530100 100 g Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BP2530500 Supplier Fisher BioReagents Supplier No. BP2530500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 617-04-9 | C7H14O6 | 194.18 g/mol

Methyl α-D-mannopyranoside, ≥99%

  • Methyl α-D-mannopyranoside is a lectin inhibitory sugar
  • Competitive inhibitor of the binding of mannose by E. coli
  • Intended for research use only

Chemical Identifiers

CAS 617-04-9
Molecular Formula C7H14O6
Molecular Weight (g/mol) 194.18
MDL Number MFCD00063262
InChI Key HOVAGTYPODGVJG-UHFFFAOYNA-N
Synonym methyl a-d-mannopyranoside, methyl alpha-d-mannopyranoside, methyl alpha-d-mannoside, alpha-methyl-d-mannoside, alpha-methyl mannopyranoside, 1-o-methyl-alpha-d-mannopyranoside, methyl-alpha-d-mannoside, alpha-d-methyl mannoside, o1-methyl-mannose, alpha-methyl-d---mannoside
PubChem CID 101798
ChEBI CHEBI:43943
SMILES COC1OC(CO)C(O)C(O)C1O

Specifications

Melting Point 193°C to 196°C
Color White
Loss on Drying < 0.5%
Assay Percent Range ≥99%
Packaging Amber Glass
Quantity 500 g
Specific Rotation 76 to 85° (+ or -)
IUPAC Name (2R,3S,4S,5S,6S)-2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol
Formula Weight 194.2
Percent Purity ≥99%
Physical Form Powder or Solid
Chemical Name or Material Methyl α-D-Mannopyranoside
Show More Show Less
Safety handling Check Alert storage code Red
Recommended Storage RT