missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl D-(-)-Mandelate 98.0+%, TCI America™
$32.72 - $48.13
Chemical Identifiers
| CAS | 20698-91-3 |
|---|---|
| Molecular Formula | C9H10O3 |
| Molecular Weight (g/mol) | 166.176 |
| MDL Number | MFCD00064247 |
| InChI Key | ITATYELQCJRCCK-MRVPVSSYSA-N |
| Synonym | methyl r---mandelate, r---methyl mandelate, r-methyl 2-hydroxy-2-phenylacetate, d---mandelic acid methyl ester, methyl 2r-2-hydroxy-2-phenylacetate, r-methyl mandelate, methyl d---mandelate, r-mandelic acid methyl ester, r---mandelic acid methyl ester, methyl hydroxy phenyl acetate # |
| PubChem CID | 2724623 |
| IUPAC Name | methyl (2R)-2-hydroxy-2-phenylacetate |
| SMILES | COC(=O)C(C1=CC=CC=C1)O |
Chemical Identifiers
| 20698-91-3 | |
| 166.176 | |
| ITATYELQCJRCCK-MRVPVSSYSA-N | |
| 2724623 | |
| COC(=O)C(C1=CC=CC=C1)O |
| C9H10O3 | |
| MFCD00064247 | |
| methyl r---mandelate, r---methyl mandelate, r-methyl 2-hydroxy-2-phenylacetate, d---mandelic acid methyl ester, methyl 2r-2-hydroxy-2-phenylacetate, r-methyl mandelate, methyl d---mandelate, r-mandelic acid methyl ester, r---mandelic acid methyl ester, methyl hydroxy phenyl acetate # | |
| methyl (2R)-2-hydroxy-2-phenylacetate |
Specifications
| 20698-91-3 | |
| C9H10O3 | |
| 1 g | |
| ITATYELQCJRCCK-MRVPVSSYSA-N | |
| methyl (2R)-2-hydroxy-2-phenylacetate | |
| 2724623 | |
| ≥98.0% (GC) | |
| Methyl D-(-)-Mandelate |
| White | |
| MFCD00064247 | |
| methyl r---mandelate, r---methyl mandelate, r-methyl 2-hydroxy-2-phenylacetate, d---mandelic acid methyl ester, methyl 2r-2-hydroxy-2-phenylacetate, r-methyl mandelate, methyl d---mandelate, r-mandelic acid methyl ester, r---mandelic acid methyl ester, methyl hydroxy phenyl acetate # | |
| COC(=O)C(C1=CC=CC=C1)O | |
| 166.176 | |
| 166.18 | |
| Crystalline Powder |
Safety and Handling
TSCA : No