missing translation for 'onlineSavingsMsg'
Learn More

Methyl D-3-Phenyllactate 98.0+%, TCI America™

Catalog No. p-7133176
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M23501G 1 g
M23505G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M23501G Supplier TCI America Supplier No. M23501G
Only null left

Chemical Identifiers

CAS 27000-00-6
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.20
MDL Number MFCD09842268
InChI Key NMPPJJIBQQCOOI-UHFFFAOYNA-N
Synonym D-3-Phenyllactic Acid Methyl Ester, Methyl (R)-2-Hydroxy-3-phenylpropionate, (R)-2-Hydroxy-3-phenylpropionic Acid Methyl Ester
PubChem CID 10986876
IUPAC Name methyl 2-hydroxy-3-phenylpropanoate
SMILES COC(=O)C(O)CC1=CC=CC=C1

Specifications

Melting Point 49°C
Color White
Boiling Point 115°C
Quantity 1 g
Formula Weight 180.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl D-3-Phenyllactate