Learn More
Methyl cyanoacetate, 99%
CAS: 105-34-0 | C4H5NO2 | 99.09 g/mol
Supplier: Thermo Scientific Chemicals A155550B
Description
Methyl cyanoacetate is an intermediate in the preparation of α-cyanoacrylate, malononitrile, 1,1-cyclohexanediacetic acid dye. It is used as UV absorber or stabiliser, intermediate for pharmaceuticals. It may be used in the synthesis of various 1,2,5-tricarbonyl compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethyl cyanoacetate is an intermediate in the preparation of α-cyanoacrylate, malononitrile, 1,1-cyclohexanediacetic acid dye. It is used as UV absorber or stabiliser, intermediate for pharmaceuticals. It may be used in the synthesis of various 1,2,5-tricarbonyl compounds.
Solubility
Soluble in water 117 g/L.
Notes
Light sensitive. Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
Specifications
| Methyl cyanoacetate | |
| -23°C | |
| 204°C | |
| Mild | |
| 1.4185 | |
| 1000 g | |
| methyl cyanoacetate, cyanoacetic acid methyl ester, methyl cyanoethanoate, acetic acid, cyano-, methyl ester, usaf kf-22, methylester kyseliny kyanoctove, methylcyanoacetate, malonic methyl ester nitrile, acetic acid, 2-cyano-, methyl ester, methyl cyano acetate | |
| ANGDWNBGPBMQHW-UHFFFAOYSA-N | |
| methyl 2-cyanoacetate | |
| 7747 | |
| 99.09 |
| 105-34-0 | |
| 1.127 | |
| 110°C (230°F) | |
| C4H5NO2 | |
| MFCD00001939 | |
| 773945 | |
| Soluble in water 117g/L. | |
| COC(=O)CC#N | |
| 99.09 | |
| CHEBI:51928 | |
| 99% |
Chemical Identifiers
| 105-34-0 | |
| 99.09 | |
| ANGDWNBGPBMQHW-UHFFFAOYSA-N | |
| 7747 | |
| methyl 2-cyanoacetate |
| C4H5NO2 | |
| MFCD00001939 | |
| methyl cyanoacetate, cyanoacetic acid methyl ester, methyl cyanoethanoate, acetic acid, cyano-, methyl ester, usaf kf-22, methylester kyseliny kyanoctove, methylcyanoacetate, malonic methyl ester nitrile, acetic acid, 2-cyano-, methyl ester, methyl cyano acetate | |
| CHEBI:51928 | |
| COC(=O)CC#N |
Safety and Handling
GHS H Statement
H319
Causes serious eye irritation.
GHS P Statement
P261-P305+P351+P338
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Warning
EINECSNumber : 203-288-8
RTECSNumber : AG4375000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only