missing translation for 'onlineSavingsMsg'
Learn More

Methyl Coumalate 96.0+%, TCI America™

Catalog No. C17345G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C17345G 5 g
C173425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C17345G Supplier TCI America Supplier No. C17345G

Chemical Identifiers

CAS 6018-41-3
Molecular Formula C7H6O4
Molecular Weight (g/mol) 154.121
MDL Number MFCD00010120
InChI Key HHWWWZQYHPFCBY-UHFFFAOYSA-N
Synonym methyl coumalate, methyl 2-oxo-2h-pyran-5-carboxylate, coumalic acid, methyl ester, 5-carbomethoxy-2-pyrone, 2h-pyran-5-carboxylic acid, 2-oxo-, methyl ester, coumalic acid methyl ester, unii-1s7tmf0r8t, methyl 2-pyrone-5-carboxylate, methyl 2-oxopyran-5-carboxylate, 1s7tmf0r8t
PubChem CID 80113
IUPAC Name methyl 6-oxopyran-3-carboxylate
SMILES COC(=O)C1=COC(=O)C=C1

Specifications

Melting Point 68°C
Color White-Yellow
Boiling Point 260°C
Quantity 5 g
Formula Weight 154.12
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl Coumalate