missing translation for 'onlineSavingsMsg'
Learn More

Methyl coumalate, 98%

Catalog No. AAB2533004
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2533004 2 g
AAB2533009 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2533004 Supplier Thermo Scientific Chemicals Supplier No. B2533004
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6018-41-3 | C7H6O4 | 154.121 g/mol

Methyl coumalate is a 2-pyrone and acts as dienophile in Diels-Alder reaction. It reacts with 1,3-butadienes at 100°C to yield tetrahydrocoumarins and 4-methoxycarbonyltricyclo[3.2.1.02,7]octenes. It also reacts with cyclohexadiene to afford tetrahydronaphthalene-2-carboxylate. It undergoes Diels-Alder reaction with unactivated alkenes to afford para-substituted adducts. It has been used as reagent in phosphine-catalyzed [4+3] annulation of modified allylic carbonates. It was also used in the preparation of 7-carboxyquinolizinium derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl coumalate is a 2-pyrone and acts as dienophile in Diels-Alder reaction. It reacts with 1,3-butadienes at 100°C to yield tetrahydrocoumarins and 4-methoxycarbonyltricyclo[3.2.1.02,7]octenes. It also reacts with cyclohexadiene to afford tetrahydronaphthalene-2-carboxylate. It undergoes Diels-Alder reaction with unactivated alkenes to afford para-substituted adducts. It has been used as reagent in phosphine-catalyzed [4+3] annulation of modified allylic carbonates. It was also used in the preparation of 7-carboxyquinolizinium derivatives.

Solubility
Freely soluble in dichloromethane.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 6018-41-3
Molecular Formula C7H6O4
Molecular Weight (g/mol) 154.121
MDL Number MFCD00010120
InChI Key HHWWWZQYHPFCBY-UHFFFAOYSA-N
Synonym methyl coumalate, methyl 2-oxo-2h-pyran-5-carboxylate, coumalic acid, methyl ester, 5-carbomethoxy-2-pyrone, 2h-pyran-5-carboxylic acid, 2-oxo-, methyl ester, coumalic acid methyl ester, unii-1s7tmf0r8t, methyl 2-pyrone-5-carboxylate, methyl 2-oxopyran-5-carboxylate, 1s7tmf0r8t
PubChem CID 80113
IUPAC Name methyl 6-oxopyran-3-carboxylate
SMILES COC(=O)C1=COC(=O)C=C1

Specifications

Melting Point 68°C to 72°C
Boiling Point 178°C to 180°C (60 mmHg)
Quantity 2 g
Merck Index 14,2557
Solubility Information Freely soluble in dichloromethane.
Formula Weight 154.12
Percent Purity 98%
Chemical Name or Material Methyl coumalate
EINECSNumber 227-871-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only