missing translation for 'onlineSavingsMsg'
Learn More

Methyl chloroformate, 99%

Catalog No. AC126531000
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC126531000 100 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC126531000 Supplier Thermo Scientific Chemicals Supplier No. 126531000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 79-22-1 | C2H3ClO2 | 94.49 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent

Chemical Identifiers

CAS 79-22-1 , 75-44-5 , 67-56-1
Molecular Formula C2H3ClO2
Molecular Weight (g/mol) 94.49
MDL Number MFCD00000639
InChI Key XMJHPCRAQCTCFT-UHFFFAOYSA-N
Synonym methyl chloroformate, methyl chlorocarbonate, chloroformic acid methyl ester, methoxycarbonyl chloride, carbonochloridic acid, methyl ester, methyl chloridocarbonate, chloro methoxy methanone, metilcloroformiato, k-stoff, methylchloorformiaat
PubChem CID 6586
IUPAC Name methyl carbonochloridate
SMILES COC(Cl)=O

Specifications

Melting Point -61.0°C
Color Colorless to Yellow
Density 1.2230g/mL
Boiling Point 70.0°C to 72.0°C
Flash Point 5°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.3860 to 1.3880
Linear Formula ClCO2CH3
Quantity 100 mL
Beilstein 03, 9
Fieser 06,376; 07,236; 09,306; 11,336
Specific Gravity 1.223
Merck Index 15, 6114
Solubility Information Solubility in water: hydrolysis. Other solubilities: miscible with alcohol, methanol, benzene,, chloroform and ether
Viscosity 0.48 mPa.s (20°C)
Formula Weight 94.5
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Methyl chloroformate, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Fatal if swallowed.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Rinse mouth.
Immediately call a POISON CENT
EINECSNumber 201-187-3
Recommended Storage Refrigerator +4°C

RUO – Research Use Only