missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Carbamate 98.0+%, TCI America™
$36.07 - $93.03
Chemical Identifiers
| CAS | 598-55-0 |
|---|---|
| Molecular Formula | C2H5NO2 |
| Molecular Weight (g/mol) | 75.07 |
| MDL Number | MFCD00007964 |
| InChI Key | GTCAXTIRRLKXRU-UHFFFAOYSA-N |
| Synonym | urethylane, carbamic acid, methyl ester, methylurethan, carbamic acid methyl ester, methylkarbamat, methylkarbamat czech, unii-9wfx634x2t, ccris 885, methylester kyseliny karbaminove, methylester kyseliny karbaminove czech |
| PubChem CID | 11722 |
| ChEBI | CHEBI:76606 |
| IUPAC Name | methyl carbamate |
| SMILES | COC(N)=O |
Chemical Identifiers
| 598-55-0 | |
| 75.07 | |
| GTCAXTIRRLKXRU-UHFFFAOYSA-N | |
| 11722 | |
| methyl carbamate |
| C2H5NO2 | |
| MFCD00007964 | |
| urethylane, carbamic acid, methyl ester, methylurethan, carbamic acid methyl ester, methylkarbamat, methylkarbamat czech, unii-9wfx634x2t, ccris 885, methylester kyseliny karbaminove, methylester kyseliny karbaminove czech | |
| CHEBI:76606 | |
| COC(N)=O |
Specifications
| 54°C | |
| 177°C | |
| C2H5NO2 | |
| urethylane, carbamic acid, methyl ester, methylurethan, carbamic acid methyl ester, methylkarbamat, methylkarbamat czech, unii-9wfx634x2t, ccris 885, methylester kyseliny karbaminove, methylester kyseliny karbaminove czech | |
| COC(N)=O | |
| 25 g | |
| 11722 | |
| 75.07 | |
| Crystalline Powder |
| 598-55-0 | |
| White | |
| MFCD00007964 | |
| GTCAXTIRRLKXRU-UHFFFAOYSA-N | |
| methyl carbamate | |
| 75.07 | |
| CHEBI:76606 | |
| ≥98.0% (GC) | |
| Methyl Carbamate |
Safety and Handling
RTECSNumber : FC2450000
TSCA : Yes