missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Calcein Blue Hydrate [Indicator for complexometry Copper], TCI America™
Specifications
Specifications
| Quantity | 1 g |
| Formula Weight | 277.28 |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Methyl Calcein Blue Hydrate [Indicator for complexometry Copper] |
Safety and Handling
| TSCA | No |