missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Bromoacetate 98.0+%, TCI America™
$52.19 - $316.89
Chemical Identifiers
| CAS | 96-32-2 |
|---|---|
| Molecular Formula | C3H5BrO2 |
| Molecular Weight (g/mol) | 152.975 |
| MDL Number | MFCD00000189 |
| InChI Key | YDCHPLOFQATIDS-UHFFFAOYSA-N |
| Synonym | methyl bromoacetate, bromoacetic acid methyl ester, methyl monobromoacetate, acetic acid, bromo-, methyl ester, methyl alpha-bromoacetate, methylbromoacetate, acetic acid, 2-bromo-, methyl ester, methylester kyseliny bromoctove, methyl .alpha.-bromoacetate, methylester kyseliny bromoctove czech |
| PubChem CID | 60984 |
| IUPAC Name | methyl 2-bromoacetate |
| SMILES | COC(=O)CBr |
Chemical Identifiers
| 96-32-2 | |
| 152.975 | |
| YDCHPLOFQATIDS-UHFFFAOYSA-N | |
| 60984 | |
| COC(=O)CBr |
| C3H5BrO2 | |
| MFCD00000189 | |
| methyl bromoacetate, bromoacetic acid methyl ester, methyl monobromoacetate, acetic acid, bromo-, methyl ester, methyl alpha-bromoacetate, methylbromoacetate, acetic acid, 2-bromo-, methyl ester, methylester kyseliny bromoctove, methyl .alpha.-bromoacetate, methylester kyseliny bromoctove czech | |
| methyl 2-bromoacetate |
Specifications
| 96-32-2 | |
| 144°C | |
| MFCD00000189 | |
| 2643 | |
| YDCHPLOFQATIDS-UHFFFAOYSA-N | |
| methyl 2-bromoacetate | |
| 60984 | |
| ≥98.0% (GC) | |
| Methyl Bromoacetate |
| Yellow | |
| C3H5BrO2 | |
| 25 g | |
| methyl bromoacetate, bromoacetic acid methyl ester, methyl monobromoacetate, acetic acid, bromo-, methyl ester, methyl alpha-bromoacetate, methylbromoacetate, acetic acid, 2-bromo-, methyl ester, methylester kyseliny bromoctove, methyl .alpha.-bromoacetate, methylester kyseliny bromoctove czech | |
| COC(=O)CBr | |
| 152.975 | |
| 152.98 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-2635&(2)-1179
RTECSNumber : AF6300000
TSCA : Yes