missing translation for 'onlineSavingsMsg'
Learn More

Methyl Bromoacetate 98.0+%, TCI America™

Catalog No. B053325G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B053325G 25 g
B0533100G 100 g
B0533500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B053325G Supplier TCI America Supplier No. B053325G

Chemical Identifiers

CAS 96-32-2
Molecular Formula C3H5BrO2
Molecular Weight (g/mol) 152.975
MDL Number MFCD00000189
InChI Key YDCHPLOFQATIDS-UHFFFAOYSA-N
Synonym methyl bromoacetate, bromoacetic acid methyl ester, methyl monobromoacetate, acetic acid, bromo-, methyl ester, methyl alpha-bromoacetate, methylbromoacetate, acetic acid, 2-bromo-, methyl ester, methylester kyseliny bromoctove, methyl .alpha.-bromoacetate, methylester kyseliny bromoctove czech
PubChem CID 60984
IUPAC Name methyl 2-bromoacetate
SMILES COC(=O)CBr

Specifications

Color Yellow
Boiling Point 144°C
Quantity 25 g
UN Number 2643
Formula Weight 152.98
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Bromoacetate
EINECSNumber (2)-2635&(2)-1179
RTECSNumber AF6300000
TSCA Yes