missing translation for 'onlineSavingsMsg'
Learn More

Methyl bromoacetate, 99%

Catalog No. AC169550250
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC169550250 25 g Glass bottle
AC169551000 100 g Glass bottle
AC169555000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC169550250 Supplier Thermo Scientific Chemicals Supplier No. 169550250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 96-32-2 | C3H5BrO2 | 152.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 96-32-2
Molecular Formula C3H5BrO2
Molecular Weight (g/mol) 152.98
MDL Number MFCD00000189
InChI Key YDCHPLOFQATIDS-UHFFFAOYSA-N
Synonym methyl bromoacetate, bromoacetic acid methyl ester, methyl monobromoacetate, acetic acid, bromo-, methyl ester, methyl alpha-bromoacetate, methylbromoacetate, acetic acid, 2-bromo-, methyl ester, methylester kyseliny bromoctove, methyl .alpha.-bromoacetate, methylester kyseliny bromoctove czech
PubChem CID 60984
IUPAC Name methyl 2-bromoacetate
SMILES COC(=O)CBr

Specifications

Color Colorless to Yellow
Density 1.6100g/mL
Boiling Point 145.0°C
Flash Point 62°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.4570 to 1.4590
Linear Formula BrCH2CO2CH3
Quantity 25 g
Beilstein 02, 213
Specific Gravity 1.61
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in methanol, ether and acetone
Formula Weight 152.98
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Methyl bromoacetate, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
EINECSNumber 202-499-2
RTECSNumber AF6300000
TSCA TSCA

RUO – Research Use Only