missing translation for 'onlineSavingsMsg'
Learn More

Methyl biphenyl-4-carboxylate, 98+%

Catalog No. AAA1343006
Change view
Click to view available options
Quantity:
5 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1343006 5 g
AAA1343018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1343006 Supplier Thermo Scientific Chemicals Supplier No. A1343006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 720-75-2 | C14H12O2 | 212.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 720-75-2
Molecular Formula C14H12O2
Molecular Weight (g/mol) 212.25
MDL Number MFCD00017200
InChI Key GATUGNVDXMYTJX-UHFFFAOYSA-N
Synonym methyl 4-biphenylcarboxylate, methyl biphenyl-4-carboxylate, methyl 1,1'-biphenyl-4-carboxylate, methyl p-phenylbenzoate, 4-biphenylcarboxylic acid, methyl ester, 1,1'-biphenyl-4-carboxylic acid, methyl ester, p-phenylbenzoic acid methyl ester, methyl-4-phenylbenzoate, 4-phenylbenzoic acid methyl ester
PubChem CID 69757
IUPAC Name methyl 4-phenylbenzoate
SMILES COC(=O)C1=CC=C(C=C1)C1=CC=CC=C1

Specifications

Melting Point 116°C to 119°C
Odor Odorless
Quantity 5 g
Beilstein 1954046
Formula Weight 212.25
Percent Purity ≥98%
Chemical Name or Material Methyl biphenyl-4-carboxylate
EINECSNumber 211-954-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only