missing translation for 'onlineSavingsMsg'
Learn More

Methyl beta-L-arabinopyranoside, 98+%

Catalog No. AAL1974603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1974603 1 g
AAL1974606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1974603 Supplier Thermo Scientific Chemicals Supplier No. L1974603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1825-00-9 | C6H12O5 | 164.157 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1825-00-9
Molecular Formula C6H12O5
Molecular Weight (g/mol) 164.157
MDL Number MFCD00047547
InChI Key ZBDGHWFPLXXWRD-AZGQCCRYSA-N
Synonym methyl beta-l-arabinopyranoside, 2s,3r,4s,5s-2-methoxytetrahydro-2h-pyran-3,4,5-triol, 2s,3r,4s,5s-2-methoxyoxane-3,4,5-triol, methyl-b-l-arabinopyranoside, methyl, a-l-arabinopyranoside, methyl-beta-l-arabinopyranoside, .beta.-l-arabinopyranoside, methyl, methyl .beta.-l-arabinopyranoside, .beta.-l-arabinopyranose methyl glycoside, arabinopyranoside, methyl, .beta.-l
PubChem CID 102169
IUPAC Name (2S,3R,4S,5S)-2-methoxyoxane-3,4,5-triol
SMILES COC1C(C(C(CO1)O)O)O

Specifications

Melting Point 167°C to 172°C
Quantity 1 g
Beilstein 4658388
Optical Rotation +243° (c=2 in Water)
Formula Weight 164.16
Percent Purity ≥98%
Chemical Name or Material Methyl beta-L-arabinopyranoside
EINECSNumber 217-362-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only