missing translation for 'onlineSavingsMsg'
Learn More

Methyl beta-D-Ribofuranoside 97.0+%, TCI America™

Catalog No. p-7132942
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M19651G 1 g
M19655G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M19651G Supplier TCI America Supplier No. M19651G
Only null left

Chemical Identifiers

CAS 7473-45-2
Molecular Formula C6H12O5
Molecular Weight (g/mol) 164.157
MDL Number MFCD00047075
InChI Key NALRCAPFICWVAQ-KVTDHHQDSA-N
Synonym methyl beta-d-ribofuranoside, methyl b-d-ribofuranoside, methyl, a-d-ribofuranoside, 2r,3s,4r,5r-2-hydroxymethyl-5-methoxytetrahydrofuran-3,4-diol, 2r,3s,4r,5r-2-hydroxymethyl-5-methoxyoxolane-3,4-diol, methyl pentofuranoside #, methyl .beta.-d-ribofuranoside, 1-o-methyl-beta-d-ribofuranoside, pubchem10614, methyl-b-d-ribofuranoside
PubChem CID 81983
IUPAC Name (2R,3S,4R,5R)-2-(hydroxymethyl)-5-methoxyoxolane-3,4-diol
SMILES COC1C(C(C(O1)CO)O)O

Specifications

Melting Point 78°C
Color White
Quantity 1 g
Formula Weight 164.16
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl beta-D-Ribofuranoside