missing translation for 'onlineSavingsMsg'
Learn More

Methyl beta-D-Glucopyranoside Hemihydrate 98.0+%, TCI America™

Supplier:  TCI America M070925G

 View more versions of this product

Catalog No. M070925G


Only null left

Description

Description

Specifications

Specifications

Methyl beta-D-Glucopyranoside Hemihydrate
111°C
C7H14O6
25 g
HOVAGTYPODGVJG-UHFFFAOYNA-N
2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol
445238
194.18
Crystalline Powder
709-50-2
White
MFCD00006602
Methyl beta-D-Glucoside
COC1OC(CO)C(O)C(O)C1O
194.18
CHEBI:320055
≥98.0% (GC)

Chemical Identifiers

709-50-2
194.18
HOVAGTYPODGVJG-UHFFFAOYNA-N
445238
2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol
C7H14O6
MFCD00006602
Methyl beta-D-Glucoside
CHEBI:320055
COC1OC(CO)C(O)C(O)C1O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions